Assay Details
Amplicon Length: | 89 |
Chromosome Location: | Chr. 5 - 77272600 - 77272600 [-] on 77272600 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-5098 | MIMAT0020605 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 89 |
Chromosome Location: | Chr. 5 - 77272600 - 77272600 [-] on 77272600 Build GRCm38 |
Species: | Mouse |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-5098 | MIMAT0020605 |