Assay Details
Gene Family ID: | MIPF0000551, mir-554 |
Mature miRNA Sequence: | GCUAGUCCUGACUCAGCCAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-554 | MIMAT0003217 |
| Pan troglodytes | ptr-miR-554 | MIMAT0008235 |
| Rhesus monkey | mml-miR-554 | MIMAT0006423 |
Stem-loop Details
Human
Pan troglodytes
Rhesus monkey

Genomic Map