Assay Details
Mature miRNA Sequence: | CAUUCUCGUUUCCUUCCCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-698-3p | MIMAT0003488 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-698-3p |
---|---|
miRBase Accession Number: | MI0004682 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CAUUCUCGUUUCCUUCCCU |
Species: | Mouse |
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | CAUUCUCGUUUCCUUCCCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-698-3p | MIMAT0003488 |