Assay Details
Gene Family ID: | MIPF0000624, mir-762 |
Mature miRNA Sequence: | GGGGCUGGGGCCGGGACAGAGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-762 | MIMAT0003892 |
Stem-loop Details
Mouse

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | mmu-miR-762 |
---|---|
miRBase Accession Number: | MI0004215 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GGGGCUGGGGCCGGGACAGAGC |
Species: | Mouse |
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000624, mir-762 |
Mature miRNA Sequence: | GGGGCUGGGGCCGGGACAGAGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-762 | MIMAT0003892 |