Assay Details
Gene Family ID: | MIPF0000391, mir-672 |
Mature miRNA Sequence: | UGAGGUUGGUGUACUGUGUGUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-672-5p | MIMAT0003735 |
| Rat | rno-miR-672-5p | MIMAT0005327 |
| Cricetulus griseus | cgr-miR-672 | MIMAT0023998 |
| Dasypus novemcinctus | dno-miR-672-5p | MIMAT0048000 |
| Horse | eca-miR-672 | MIMAT0013242 |
| Pteropus alecto | pal-miR-672-5p | MIMAT0039813 |
| View More Rows | ||
| Rabbit | ocu-miR-672-5p | MIMAT0048557 |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Dasypus novemcinctus
Horse
Pteropus alecto
Rabbit
More Information
Related Products
miRBase Alias:
Human: hsa-miR-672 (v10)
Mouse: mmu-miR-672 (v17)
Rat: rno-miR-672 (v18)
Mouse: mmu-miR-672 (v17)
Rat: rno-miR-672 (v18)

Genomic Map