Assay Details
Gene Family ID: | MIPF0000063, mir-192 |
Mature miRNA Sequence: | AUGACCUAUGAUUUGACAGACA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-215 | MIMAT0003118 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | rno-miR-215 |
---|---|
miRBase Accession Number: | MI0003482 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AUGACCUAUGAUUUGACAGACA |
Species: | Rat |
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000063, mir-192 |
Mature miRNA Sequence: | AUGACCUAUGAUUUGACAGACA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-215 | MIMAT0003118 |