Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGGUUGUCUGUGAUGAGUUCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-410-5p | MIMAT0026558 |
| Mouse | mmu-miR-410-5p | MIMAT0017172 |
| Rat | rno-miR-410-5p | MIMAT0017303 |
| Cricetulus griseus | cgr-miR-410-5p | MIMAT0023952 |
| Guinea pig | cpo-miR-410-5p | MIMAT0047235 |
| Pteropus alecto | pal-miR-410-5p | MIMAT0040151 |
| View More Rows | ||
| Rabbit | ocu-miR-410-5p | MIMAT0048435 |
| Rhesus monkey | mml-miR-410-5p | MIMAT0026869 |
Stem-loop Details
Human
Mouse
Rat
Cricetulus griseus
Guinea pig
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-410* (v17)
Rat: rno-miR-410* (v18)
Rat: rno-miR-410* (v18)

Genomic Map