Assay Details
Amplicon Length: | 81 |
Chromosome Location: | Chr. 4 - 28659377 - 28659377 [-] on 28659377 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-489-5p | MIMAT0017196 |
| Rat | rno-miR-489-3p | MIMAT0003113 |
| Rat | rno-miR-653-5p | MIMAT0012838 |
| Rat | rno-miR-653-3p | MIMAT0017361 |
Stem-loop Details
Rat

Genomic Map