Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 8 - 45746794 - 45746794 [+] on 45746794 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-100-5p | MIMAT0000822 |
| Rat | rno-miR-100-3p | MIMAT0017112 |
Stem-loop Details
Rat

Genomic Map