Assay Details
Amplicon Length: | 96 |
Chromosome Location: | Chr. X - 151253063 - 151253063 [+] on 151253063 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-743b-3p | MIMAT0005280 |
Rat | rno-miR-743b-5p | MIMAT0017288 |
Stem-loop Details
Rat
