Assay Details
Amplicon Length: | 78 |
Chromosome Location: | Chr. 1 - 240403554 - 240403554 [+] on 240403554 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-204-5p | MIMAT0000877 |
| Rat | rno-miR-204-3p | MIMAT0004739 |
Stem-loop Details
Rat

Genomic Map