Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. X - 155338668 - 155338668 [-] on 155338668 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-547-3p | MIMAT0012851 |
Rat | rno-miR-547-5p | MIMAT0017368 |
Stem-loop Details
Rat
