Assay Details
Amplicon Length: | 112 |
Chromosome Location: | Chr. X - 155344047 - 155344047 [-] on 155344047 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3585-5p | MIMAT0017878 |
Rat | rno-miR-3585-3p | MIMAT0017879 |
Stem-loop Details
Rat
