Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAUGUAACACGGUCCACUAACC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-411-3p | MIMAT0004813 |
Mouse | mmu-miR-411-3p | MIMAT0001093 |
Stem-loop Details
Human
Mouse
