Assay Details
| Gene Family ID: | MIPF0000065, mir-132 | 
| Mature miRNA Sequence: | UAACAGUCUACAGCCAUGGUCG | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-132-3p | MIMAT0000426 | 
| Mouse | mmu-miR-132-3p | MIMAT0000144 | 
| Rat | rno-miR-132-3p | MIMAT0000838 | 
| Alligator mississippiensis | ami-miR-132-3p | MIMAT0038145 | 
| Bovine | bta-miR-132 | MIMAT0003812 | 
| Callithrix jacchus | cja-miR-132 | MIMAT0039423 | 
| View More Rows | ||
| Chicken | gga-miR-132a-3p | MIMAT0049951 | 
| Chrysemys picta | cpi-miR-132-3p | MIMAT0037749 | 
| Cricetulus griseus | cgr-miR-132-3p | MIMAT0023759 | 
| Cyprinus carpio | ccr-miR-132a | MIMAT0026213 | 
| Dasypus novemcinctus | dno-miR-132-3p | MIMAT0047632 | 
| Eptesicus fuscus | efu-miR-132 | MIMAT0035002 | 
| Fugu rubripes | fru-miR-132 | MIMAT0003059 | 
| Guinea pig | cpo-miR-132-3p | MIMAT0046985 | 
| Horse | eca-miR-132 | MIMAT0013021 | 
| Ictalurus punctatus | ipu-miR-132b | MIMAT0029406 | 
| Microcebus murinus | mmr-miR-132 | MIMAT0049229 | 
| Monodelphis domestica | mdo-miR-132-3p | MIMAT0004149 | 
| Otolemur garnettii | oga-miR-132 | MIMAT0049698 | 
| Pig | ssc-miR-132 | MIMAT0025361 | 
| Pongo pygmaeus | ppy-miR-132 | MIMAT0015755 | 
| Pteropus alecto | pal-miR-132-3p | MIMAT0039992 | 
| Rhesus monkey | mml-miR-132-3p | MIMAT0006188 | 
| Salmo salar | ssa-miR-132-3p | MIMAT0032332 | 
| Taeniopygia guttata | tgu-miR-132 | MIMAT0018619 | 
| Tetraodon nigroviridis | tni-miR-132 | MIMAT0003060 | 
| Xenopus tropicalis | xtr-miR-132 | MIMAT0003594 | 
| Zebrafish | dre-miR-132-3p | MIMAT0001829 | 
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-132 | 
| Stem-loop Accession # | MI0000449 | 
| Stem-loop Sequence | CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC | 
| Chromosome Location | Chr. 17 - 2049908 - 2050008 [-] on Build GRCh38 | 
| Mature MicroRNA | miRBase Accession #: MIMAT0000426 miRBase ID: hsa-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: Chr. 17 - 2049908 - 2050008 [-] on Build GRCh38 | 
| Stem-loop ID | mmu-mir-132 | 
| Stem-loop Accession # | MI0000158 | 
| Stem-loop Sequence | GGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCC | 
| Chromosome Location | Chr. 11 - 75173682 - 75173747 [+] on Build GRCm38 | 
| Mature MicroRNA | miRBase Accession #: MIMAT0000144 miRBase ID: mmu-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: Chr. 11 - 75173682 - 75173747 [+] on Build GRCm38 | 
| Stem-loop ID | rno-mir-132 | 
| Stem-loop Accession # | MI0000905 | 
| Stem-loop Sequence | CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCUC | 
| Chromosome Location | Chr. 10 - 62014995 - 62015095 [+] on Build Rnor_6.0 | 
| Mature MicroRNA | miRBase Accession #: MIMAT0000838 miRBase ID: rno-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: Chr. 10 - 62014995 - 62015095 [+] on Build Rnor_6.0 | 
| Stem-loop ID | ami-mir-132 | 
| Stem-loop Accession # | MI0029732 | 
| Stem-loop Sequence | ACGUCGCCAGGGCAACCGUGGCUUUAGAUUGUUACUGUGUGGUGCGUGGGUAACAGUCUACAGCCAUGGUCGCUGGGCCGGACGC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0038145 miRBase ID: ami-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | bta-mir-132 | 
| Stem-loop Accession # | MI0005028 | 
| Stem-loop Sequence | CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003812 miRBase ID: bta-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | cja-mir-132 | 
| Stem-loop Accession # | MI0031942 | 
| Stem-loop Sequence | GGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0039423 miRBase ID: cja-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | gga-mir-132a | 
| Stem-loop Accession # | MI0040692 | 
| Stem-loop Sequence | ACCGUGGCUUUAGAUUGUUACUGUGCUGCUGGGGGGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049951 miRBase ID: gga-miR-132a-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | cpi-mir-132 | 
| Stem-loop Accession # | MI0029471 | 
| Stem-loop Sequence | UGUCUCCAGGGCAACCGUGGCUUUAGAUUGUUACUGUGGUUGUGUGUUGGUAACAGUCUACAGCCAUGGUCGCUUGGGCUAGACACAC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0037749 miRBase ID: cpi-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | cgr-mir-132 | 
| Stem-loop Accession # | MI0020396 | 
| Stem-loop Sequence | UCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0023759 miRBase ID: cgr-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | ccr-mir-132a | 
| Stem-loop Accession # | MI0023320 | 
| Stem-loop Sequence | CUUGCUGUCUCCAUGGUGACCGUGGCAUUAGAUUGUUACUGUAGCAACAGCACUUCUGGUAACAGUCUACAGCCAUGGUCGCUAGGGGGCAG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0026213 miRBase ID: ccr-miR-132a Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | dno-mir-132 | 
| Stem-loop Accession # | MI0038976 | 
| Stem-loop Sequence | ACCGUGGCUUUCGAUUGUUACUGUGGAAACCGGAGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0047632 miRBase ID: dno-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | efu-mir-132 | 
| Stem-loop Accession # | MI0028687 | 
| Stem-loop Sequence | CUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0035002 miRBase ID: efu-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | fru-mir-132 | 
| Stem-loop Accession # | MI0003399 | 
| Stem-loop Sequence | GUCUCCAUGGUUACCGUGGCAUUAGAUUGUUACUGUAGCAACUGCACCAGUGGUAACAGUCUACAGCCAUGGUCGCUAGGGGGCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003059 miRBase ID: fru-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | cpo-mir-132 | 
| Stem-loop Accession # | MI0038635 | 
| Stem-loop Sequence | ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0046985 miRBase ID: cpo-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | eca-mir-132 | 
| Stem-loop Accession # | MI0012770 | 
| Stem-loop Sequence | GGACAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0013021 miRBase ID: eca-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | ipu-mir-132b | 
| Stem-loop Accession # | MI0024492 | 
| Stem-loop Sequence | ACCGUGGCAUUAGAUUGUUACUGUAGGAACAGAAUUUUUGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0029406 miRBase ID: ipu-miR-132b Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | mmr-mir-132 | 
| Stem-loop Accession # | MI0039957 | 
| Stem-loop Sequence | ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049229 miRBase ID: mmr-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | mdo-mir-132 | 
| Stem-loop Accession # | MI0005338 | 
| Stem-loop Sequence | GGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCAGGGGUAACAGUCUACAGCCAUGGUCGCCC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0004149 miRBase ID: mdo-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | oga-mir-132 | 
| Stem-loop Accession # | MI0040482 | 
| Stem-loop Sequence | ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0049698 miRBase ID: oga-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | ssc-mir-132 | 
| Stem-loop Accession # | MI0022129 | 
| Stem-loop Sequence | CUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0025361 miRBase ID: ssc-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | ppy-mir-132 | 
| Stem-loop Accession # | MI0014824 | 
| Stem-loop Sequence | CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0015755 miRBase ID: ppy-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | pal-mir-132 | 
| Stem-loop Accession # | MI0032437 | 
| Stem-loop Sequence | ACCGUGGCUUUCGAUUGUUACUGUGGGAACCGGAGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0039992 miRBase ID: pal-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | mml-mir-132 | 
| Stem-loop Accession # | MI0007620 | 
| Stem-loop Sequence | CCGCCCCCGCGUCUCCAGGGCAACCGUGGCUUUCGAUUGUUACUGUGGGAACUGGAGGUAACAGUCUACAGCCAUGGUCGCCCCGCAGCACGCCCACGCGC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0006188 miRBase ID: mml-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | ssa-mir-132-1 | 
| Stem-loop Accession # | MI0026475 | 
| Stem-loop Sequence | ACCGUGGCUUUAGAUUGUUACUCUAGAGACAGCACCAUGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0032332 miRBase ID: ssa-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | ssa-mir-132-2 | 
| Stem-loop Accession # | MI0026476 | 
| Stem-loop Sequence | ACCGUGGCUUUAGAUUGUUACUGUAGAGACGGUAACAUGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0032332 miRBase ID: ssa-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | ssa-mir-132-3 | 
| Stem-loop Accession # | MI0026477 | 
| Stem-loop Sequence | ACCGUGGCAUUAGAUUGUUACUGUAGAAACAGCACCACUGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0032332 miRBase ID: ssa-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | ssa-mir-132-4 | 
| Stem-loop Accession # | MI0026478 | 
| Stem-loop Sequence | ACCGUGGCAUUAGAUUGUUACUGUAGCAACAGCACCACUGGUAACAGUCUACAGCCAUGGUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0032332 miRBase ID: ssa-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | tgu-mir-132 | 
| Stem-loop Accession # | MI0016249 | 
| Stem-loop Sequence | AUCCGACGCCGCCCGGGCGACCGUGGCUUUAGAUAGUUACUGUUCCACUGGGGGAGUAACAGUCUACAGCCAUGGUCGCCGGGACAGGCAGACUCG | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0018619 miRBase ID: tgu-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | tni-mir-132 | 
| Stem-loop Accession # | MI0003400 | 
| Stem-loop Sequence | GUCUCCAUGGUUACCGUGGCAUUAGAUUGUUACUGUAGCAACUGCACCAGCGGUAACAGUCUACAGCCAUGGUCGCUAGGGGGCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003060 miRBase ID: tni-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | xtr-mir-132 | 
| Stem-loop Accession # | MI0004834 | 
| Stem-loop Sequence | CUGUCUCCAGGGCAACCGUGGCUUUAGAUUGUUACUGUAGUUCUGCAUUGGUAACAGUCUACAGCCAUGGUCGCUCGGGCAAGAUGC | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0003594 miRBase ID: xtr-miR-132 Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | dre-mir-132-1 | 
| Stem-loop Accession # | MI0001989 | 
| Stem-loop Sequence | GUCUCCAUGGCGACCGUGGCAUUAGAUUGUUACUGUAGGAACAGAAUUUUUGGUAACAGUCUACAGCCAUGGUCGCUAGUGGGCA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0001829 miRBase ID: dre-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
| Stem-loop ID | dre-mir-132-2 | 
| Stem-loop Accession # | MI0001991 | 
| Stem-loop Sequence | UCCCUUCCUCUGCUGUCUCCAUGCUGACCGUGGCUUUAGAUUGUUACUGUAGCGUUGGCACGUGGUAACAGUCUACAGCCAUGGUCGCCAAGGGGCAGGUGUUACAGCGACUACUGGGA | 
| Chromosome Location | - | 
| Mature MicroRNA | miRBase Accession #: MIMAT0001829 miRBase ID: dre-miR-132-3p Mature miRNA Sequence: UAACAGUCUACAGCCAUGGUCG Chromosome Location: NA | 
 Back To Top
 Back To TopMore Information
Related Products
 TaqMan™ Pri-miRNA Assay : Rn03465057_pri
						TaqMan™ Pri-miRNA Assay : Rn03465057_pri TaqMan™ Pri-miRNA Assay : Mm03306275_pri
						TaqMan™ Pri-miRNA Assay : Mm03306275_pri TaqMan™ Pri-miRNA Assay : Hs03303111_pri
						TaqMan™ Pri-miRNA Assay : Hs03303111_pri Ambion® Anti-miR™ miRNA Inhibitor : AM10166
						Ambion® Anti-miR™ miRNA Inhibitor : AM10166 Ambion® Pre-miR™ miRNA Precursor : PM10166
						Ambion® Pre-miR™ miRNA Precursor : PM10166 TaqMan™ MicroRNA Assay : 000457
						TaqMan™ MicroRNA Assay : 000457 mirVana® miRNA inhibitor : MH10166
						mirVana® miRNA inhibitor : MH10166 mirVana® miRNA mimic : MC10166
						mirVana® miRNA mimic : MC10166 TaqMan™ Advanced miRNA Assay : 477900_mir
						TaqMan™ Advanced miRNA Assay : 477900_mir TaqMan™ Advanced miRNA Assay : rno480919_mir
						TaqMan™ Advanced miRNA Assay : rno480919_mir Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top