Assay Details
| Gene Family ID: | MIPF0001105, mir-3065 | 
| Mature miRNA Sequence: | UCAGCACCAGGAUAUUGUUGGGG | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-3065-3p | MIMAT0014837 | 
| Horse | eca-miR-3065 | MIMAT0034658 | 
Stem-loop Details
Mouse
Horse

 Genomic Map
Genomic Map