Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAGUAGACCGUAUAGCGUACG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-411-5p | MIMAT0003329 |
Mouse | mmu-miR-411-5p | MIMAT0004747 |
Rat | rno-miR-411-5p | MIMAT0005312 |
Callithrix jacchus | cja-miR-411 | MIMAT0039445 |
Dasypus novemcinctus | dno-miR-411-5p | MIMAT0047874 |
Guinea pig | cpo-miR-411-5p | MIMAT0047237 |
View More Rows | ||
Horse | eca-miR-411 | MIMAT0013154 |
Microcebus murinus | mmr-miR-411a | MIMAT0049194 |
Otolemur garnettii | oga-miR-411a | MIMAT0049676 |
Pan paniscus | ppa-miR-411a | MIMAT0049046 |
Pan troglodytes | ptr-miR-411 | MIMAT0008138 |
Papio hamadryas | pha-miR-411a | MIMAT0049579 |
Pongo pygmaeus | ppy-miR-411 | MIMAT0015878 |
Rabbit | ocu-miR-411-5p | MIMAT0048437 |
Rhesus monkey | mml-miR-411-5p | MIMAT0006317 |
Tupaia chinensis | tch-miR-411-5p | MIMAT0036490 |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Dasypus novemcinctus
Guinea pig
Horse
Microcebus murinus
Otolemur garnettii
Pan paniscus
Pan troglodytes
Papio hamadryas
Pongo pygmaeus
Rabbit
Rhesus monkey
Tupaia chinensis
