Assay Details
Gene Family ID: | MIPF0000164, mir-322 |
Mature miRNA Sequence: | AAACAUGAAGCGCUGCAACAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-322-3p | MIMAT0000549 |
| Cricetulus griseus | cgr-miR-322-3p | MIMAT0023908 |
Stem-loop Details
Mouse
Cricetulus griseus

Genomic Map