Assay Details
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | CAGUUGCUAGCUGCACUACCCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-449c-3p | MIMAT0022715 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-449c-3p |
|---|---|
| Stem-loop Accession Number: | MI0004645 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGUUGCUAGCUGCACUACCCU |
| Species: | Mouse |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | CAGUUGCUAGCUGCACUACCCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-449c-3p | MIMAT0022715 |