Assay Details
Mature miRNA Sequence: | AGUGGUCCUAAACAUUUCAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-203b-5p | MIMAT0049831 |
| Rat | rno-miR-203b-5p | MIMAT0017799 |
Stem-loop Details
Mouse
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-203b-5p |
|---|---|
| Stem-loop Accession Number: | MI0040622 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGUGGUCCUAAACAUUUCAC |
| Species: | Mouse, Rat |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | AGUGGUCCUAAACAUUUCAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-203b-5p | MIMAT0049831 |
| Rat | rno-miR-203b-5p | MIMAT0017799 |