Assay Details
| Gene Family ID: | MIPF0000106, mir-137 | 
| Mature miRNA Sequence: | UUAUUGCUUAAGAAUACGCGUAG | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-137-3p | MIMAT0000429 | 
| Mouse | mmu-miR-137-3p | MIMAT0000149 | 
| Rat | rno-miR-137-3p | MIMAT0000843 | 
| Anolis carolinensis | aca-miR-137a | MIMAT0021754 | 
| Bovine | bta-miR-137 | MIMAT0009231 | 
| Callithrix jacchus | cja-miR-137 | MIMAT0039514 | 
| View More Rows | ||
| Chrysemys picta | cpi-miR-137a-3p | MIMAT0037758 | 
| Columba livia | cli-miR-137a-3p | MIMAT0038519 | 
| Cricetulus griseus | cgr-miR-137-3p | MIMAT0023763 | 
| Cyprinus carpio | ccr-miR-137 | MIMAT0026217 | 
| Dasypus novemcinctus | dno-miR-137-3p | MIMAT0047647 | 
| Horse | eca-miR-137 | MIMAT0012953 | 
| Ictalurus punctatus | ipu-miR-137 | MIMAT0029413 | 
| Oreochromis niloticus | oni-miR-137a | MIMAT0042764 | 
| Ornithorhynchus anatinus | oan-miR-137a-3p | MIMAT0006838 | 
| Pan troglodytes | ptr-miR-137 | MIMAT0008033 | 
| Pig | ssc-miR-137 | MIMAT0025362 | 
| Pongo pygmaeus | ppy-miR-137 | MIMAT0015759 | 
| Python bivittatus | pbv-miR-137a-3p | MIMAT0038946 | 
| Rhesus monkey | mml-miR-137-3p | MIMAT0006193 | 
| Taeniopygia guttata | tgu-miR-137-3p | MIMAT0014543 | 
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Bovine
Callithrix jacchus
Chrysemys picta
Columba livia
Cricetulus griseus
Cyprinus carpio
Dasypus novemcinctus
Horse
Ictalurus punctatus
Oreochromis niloticus
Ornithorhynchus anatinus
Pan troglodytes
Pig
Pongo pygmaeus
Python bivittatus
Rhesus monkey
Taeniopygia guttata
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :