Assay Details
Gene Family ID: | MIPF0000079, mir-145 |
Mature miRNA Sequence: | GGAUUCCUGGAAAUACUGUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-145-3p | MIMAT0017131 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-145-3p |
|---|---|
| Stem-loop Accession Number: | MI0000918 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGAUUCCUGGAAAUACUGUUC |
| Species: | Rat |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000079, mir-145 |
Mature miRNA Sequence: | GGAUUCCUGGAAAUACUGUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-145-3p | MIMAT0017131 |