Assay Details
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | UGUAAACAUCCUACACUCUCAGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-30c-5p | MIMAT0000244 |
Mouse | mmu-miR-30c-5p | MIMAT0000514 |
Rat | rno-miR-30c-5p | MIMAT0000804 |
Alligator mississippiensis | ami-miR-30c-5p | MIMAT0038081 |
Anolis carolinensis | aca-miR-30c-5p | MIMAT0021924 |
Astatotilapia burtoni | abu-miR-30b | MIMAT0042071 |
View More Rows | ||
Bovine | bta-miR-30c | MIMAT0003850 |
Cricetulus griseus | cgr-miR-30c-2 | MIMAT0037458 |
Cricetulus griseus | cgr-miR-30c | MIMAT0023898 |
Goat | chi-miR-30c-5p | MIMAT0036125 |
Gorilla gorilla | ggo-miR-30c | MIMAT0024111 |
Horse | eca-miR-30c | MIMAT0012915 |
Lagothrix lagotricha | lla-miR-30c | MIMAT0002615 |
Macaca nemestrina | mne-miR-30c | MIMAT0002616 |
Microcebus murinus | mmr-miR-30c | MIMAT0049218 |
Monodelphis domestica | mdo-miR-30c-5p | MIMAT0028502 |
Ornithorhynchus anatinus | oan-miR-30c-5p | MIMAT0006911 |
Oryzias latipes | ola-miR-30c | MIMAT0022598 |
Otolemur garnettii | oga-miR-30c | MIMAT0049643 |
Pan troglodytes | ptr-miR-30c | MIMAT0002614 |
Papio hamadryas | pha-miR-30c | MIMAT0049583 |
Pig | ssc-miR-30c-5p | MIMAT0002167 |
Pongo pygmaeus | ppy-miR-30c | MIMAT0015737 |
Rhesus monkey | mml-miR-30c-5p | MIMAT0006170 |
Salmo salar | ssa-miR-30a-5p | MIMAT0032600 |
Tupaia chinensis | tch-miR-30c-5p | MIMAT0036585 |
Xenopus tropicalis | xtr-miR-30c | MIMAT0003577 |
Stem-loop Details
Human
Stem-loop ID | hsa-mir-30c-1 |
Stem-loop Accession # | MI0000736 |
Stem-loop Sequence | ACCAUGCUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA |
Chromosome Location | Chr. 1 - 40757284 - 40757372 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000244
miRBase ID: hsa-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: Chr. 6 - 71376960 - 71377031 [-] on Build GRCh38 |
Stem-loop ID | hsa-mir-30c-2 |
Stem-loop Accession # | MI0000254 |
Stem-loop Sequence | AGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUUUCU |
Chromosome Location | Chr. 6 - 71376960 - 71377031 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000244
miRBase ID: hsa-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: Chr. 6 - 71376960 - 71377031 [-] on Build GRCh38 |
Stem-loop ID | mmu-mir-30c-1 |
Stem-loop Accession # | MI0000547 |
Stem-loop Sequence | ACCAUGUUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA |
Chromosome Location | Chr. 4 - 120769534 - 120769622 [-] on Build GRCm38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000514
miRBase ID: mmu-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: Chr. 1 - 23291701 - 23291784 [+] on Build GRCm38 |
Stem-loop ID | mmu-mir-30c-2 |
Stem-loop Accession # | MI0000548 |
Stem-loop Sequence | GAGUGACAGAUAUUGUAAACAUCCUACACUCUCAGCUGUGAAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUU |
Chromosome Location | Chr. 1 - 23291701 - 23291784 [+] on Build GRCm38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000514
miRBase ID: mmu-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: Chr. 1 - 23291701 - 23291784 [+] on Build GRCm38 |
Stem-loop ID | rno-mir-30c-1 |
Stem-loop Accession # | MI0000866 |
Stem-loop Sequence | ACCAUGUUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA |
Chromosome Location | Chr. 5 - 139699877 - 139699965 [-] on Build Rnor_6.0 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000804
miRBase ID: rno-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: Chr. 9 - 29562376 - 29562459 [+] on Build Rnor_6.0 |
Stem-loop ID | rno-mir-30c-2 |
Stem-loop Accession # | MI0000871 |
Stem-loop Sequence | GAGUGACAGAUACUGUAAACAUCCUACACUCUCAGCUGUGAAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUU |
Chromosome Location | Chr. 9 - 29562376 - 29562459 [+] on Build Rnor_6.0 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000804
miRBase ID: rno-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: Chr. 9 - 29562376 - 29562459 [+] on Build Rnor_6.0 |
Stem-loop ID | ami-mir-30c-1 |
Stem-loop Accession # | MI0029692 |
Stem-loop Sequence | AUCUACCAUGCCGUAGCAUGUGUAAACAUCCUACACUCUCAGCUGUGAACUCAAGGUGGCUGGGAGAGGGUUGUUUACGCCUUCUGCCAUGGUCUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038081
miRBase ID: ami-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ami-mir-30c-2 |
Stem-loop Accession # | MI0029693 |
Stem-loop Sequence | GCUCUGAGUGACAGGUAUUGUAAACAUCCUACACUCUCAGCUGUGGAAAAUGAGGAAGCUGGGAGAAGGCUGUUUACUCUCCCUGACUUAGAUAGC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0038081
miRBase ID: ami-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | aca-mir-30c-1 |
Stem-loop Accession # | MI0018849 |
Stem-loop Sequence | CCAAGUGACAGGUAAUGUAAACAUCCUACACUCUCAGCUGUGUAGAUAAGAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUUGGAAA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0021924
miRBase ID: aca-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | aca-mir-30c-2 |
Stem-loop Accession # | MI0029351 |
Stem-loop Sequence | UAGCAGCGCAUGUAAACAUCCUACACUCUCAGCUGUGAAUUUGUGGUGGCUGGGAGAAGGUUGUUUACACCUUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0021924
miRBase ID: aca-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | abu-mir-30b |
Stem-loop Accession # | MI0033766 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGUUGUCAUGGAGCUGGGAGAGGGGUGUUUACGCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0042071
miRBase ID: abu-miR-30b Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | bta-mir-30c |
Stem-loop Accession # | MI0005064 |
Stem-loop Sequence | CAGACUGUAACCAUGCCGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCGAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGAAAACGUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003850
miRBase ID: bta-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | cgr-mir-30c-2 |
Stem-loop Accession # | MI0020486 |
Stem-loop Sequence | UGACAGAUACUGUAAACAUCCUACACUCUCAGCUGUGAAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0037458
miRBase ID: cgr-miR-30c-2 Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA miRBase Accession #: MIMAT0023898 miRBase ID: cgr-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | chi-mir-30c |
Stem-loop Accession # | MI0030733 |
Stem-loop Sequence | CAGACUGUAACCAUGCCGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCGAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGAAAACGUC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036125
miRBase ID: chi-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ggo-mir-30c |
Stem-loop Accession # | MI0020658 |
Stem-loop Sequence | GAGCACUGAGCGACAGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUUUCUGCCUUGGAAGUCAACUAAAGAGAAAUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0024111
miRBase ID: ggo-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | eca-mir-30c |
Stem-loop Accession # | MI0012671 |
Stem-loop Sequence | ACCAUGCUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0012915
miRBase ID: eca-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | eca-mir-30c-2 |
Stem-loop Accession # | MI0012849 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGGAAAGUGAGAAAGCUGGGAGAAGGCUGUUUACU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0012915
miRBase ID: eca-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | lla-mir-30c |
Stem-loop Accession # | MI0002924 |
Stem-loop Sequence | AGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUUUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002615
miRBase ID: lla-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | mne-mir-30c |
Stem-loop Accession # | MI0002925 |
Stem-loop Sequence | AGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUUUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002616
miRBase ID: mne-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | mmr-mir-30c |
Stem-loop Accession # | MI0039944 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGGCAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049218
miRBase ID: mmr-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | mdo-mir-30c-1 |
Stem-loop Accession # | MI0023768 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGGAAAGUCAGAAAGCUGGGAGAAGGCUGUUACUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0028502
miRBase ID: mdo-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | mdo-mir-30c-2 |
Stem-loop Accession # | MI0023835 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGCUGUUUACUCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0028502
miRBase ID: mdo-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | oan-mir-30c-1 |
Stem-loop Accession # | MI0024093 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUCAAGUCAAGGAGGCUGGGAGAGGGUUGUUUACUCC |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0006911
miRBase ID: oan-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | oan-mir-30c-2 |
Stem-loop Accession # | MI0006728 |
Stem-loop Sequence | CUGUUCUAAAGAGCCCUGAGUGACAGGUACUGUAAACAUCCUACACUCUCAGCUGUAGAUGUGAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUUGGAAAUCAGCCAAAGAGAACAG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0006911
miRBase ID: oan-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ola-mir-30c |
Stem-loop Accession # | MI0019480 |
Stem-loop Sequence | ACCACUCUCUGAAUCUAAUGGAAAUGUAAAUUUGUUGUCUUGUAGACUGUCUCCAGGCAUGUAAACAUCCUACACUCUCAGCUGUGUUGUCAUGGAGCUGGGAGAGGGGU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0022598
miRBase ID: ola-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | oga-mir-30c |
Stem-loop Accession # | MI0040426 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAGGCUGGGAGAAGGCUGUUUACUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049643
miRBase ID: oga-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ptr-mir-30c-1 |
Stem-loop Accession # | MI0008605 |
Stem-loop Sequence | CCAUGCUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002614
miRBase ID: ptr-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ptr-mir-30c-2 |
Stem-loop Accession # | MI0002923 |
Stem-loop Sequence | AGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUUUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002614
miRBase ID: ptr-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | pha-mir-30c |
Stem-loop Accession # | MI0040360 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0049583
miRBase ID: pha-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ssc-mir-30c-1 |
Stem-loop Accession # | MI0016618 |
Stem-loop Sequence | CGUAGUGCGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCGAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002167
miRBase ID: ssc-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ssc-mir-30c-2 |
Stem-loop Accession # | MI0002461 |
Stem-loop Sequence | GACAGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0002167
miRBase ID: ssc-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ppy-mir-30c-1 |
Stem-loop Accession # | MI0014803 |
Stem-loop Sequence | ACCAUGCUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0015737
miRBase ID: ppy-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | mml-mir-30c-1 |
Stem-loop Accession # | MI0007599 |
Stem-loop Sequence | ACCAUGCUGUAGUGUGUGUAAACAUCCUACACUCUCAGCUGUGAGCUCAAGGUGGCUGGGAGAGGGUUGUUUACUCCUUCUGCCAUGGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0006170
miRBase ID: mml-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | mml-mir-30c-2 |
Stem-loop Accession # | MI0007600 |
Stem-loop Sequence | AGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0006170
miRBase ID: mml-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ssa-mir-30a-1 |
Stem-loop Accession # | MI0026676 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCGGUGCCCUCUUUCAGCCGGGAGUGGGAUGUUUGCGCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032600
miRBase ID: ssa-miR-30a-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ssa-mir-30a-2 |
Stem-loop Accession # | MI0026677 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCGGCGCCCUCUAGUAGCCGGGAGUGGGAUGUUUGCGCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032600
miRBase ID: ssa-miR-30a-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ssa-mir-30a-3 |
Stem-loop Accession # | MI0026674 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGACAUCAUCGAGCUGGGAGAGGGGUGUUUACACU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032600
miRBase ID: ssa-miR-30a-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | ssa-mir-30a-4 |
Stem-loop Accession # | MI0026675 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGACAUCAUCGAGCUGGGAGAGGGGUGUUUACGCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0032600
miRBase ID: ssa-miR-30a-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | tch-mir-30c |
Stem-loop Accession # | MI0031251 |
Stem-loop Sequence | UGUAAACAUCCUACACUCUCAGCUGUGGAAAGUGAGAAAGCUGGGAGAAGGCUGUUUACUCU |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0036585
miRBase ID: tch-miR-30c-5p Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | xtr-mir-30c-1 |
Stem-loop Accession # | MI0004815 |
Stem-loop Sequence | ACCAUGCAGCAGUGAUUGUAAACAUCCUACACUCUCAGCUGUGAACAUAAGGUGGCUGGGAGAAGGGUGUUUACUCCCCCUGCCAUGGA |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003577
miRBase ID: xtr-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |
Stem-loop ID | xtr-mir-30c-2 |
Stem-loop Accession # | MI0004919 |
Stem-loop Sequence | GAGAGACAGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAUACAAAGCUGGGGGAAGGCUGUUUACUCUCUCUG |
Chromosome Location | - |
Mature MicroRNA |
miRBase Accession #: MIMAT0003577
miRBase ID: xtr-miR-30c Mature miRNA Sequence: UGUAAACAUCCUACACUCUCAGC Chromosome Location: NA |

More Information
Related Products













