Assay Details
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | AUAUACAUACACACAUACACA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-466b-2-3p | MIMAT0017286 |
| Rat | rno-miR-466b-4-3p | MIMAT0037262 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | rno-miR-466b-2-3p |
|---|---|
| Stem-loop Accession Number: | MI0006113 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AUAUACAUACACACAUACACA |
| Species: | Rat |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | AUAUACAUACACACAUACACA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-466b-2-3p | MIMAT0017286 |
| Rat | rno-miR-466b-4-3p | MIMAT0037262 |