Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Hydroxypropyl)-Gamma-Cyclodextrin, Spectrum™ Chemical
SDP

Synonym: 3-Fluoro-4-Nitrotoluene, 2-Fluoro-4-Methyl-1-Nitrobenzene, Benzene, 2-Fluoro-4-Methyl-1-Nitro-

$258.00 - $876.00

Chemical Identifiers

CAS 128446-34-4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-620-880
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H27585GM
5 g
Each for $258.00
Add to Cart
 
18-620-881
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
H2758100GM
100 g
Each for $876.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

128446-34-4

Specifications

128446-34-4
Amber Glass Bottle
(2-+L23558:M23574Hydroxypropyl)-Gamma-Cyclodextrin
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.