Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6,9-Trioxaundecanedioic Acid, Spectrum™ Chemical
SDP

$1032.00

Chemical Identifiers

CAS 13887-98-4
Molecular Weight (g/mol) 222.19 g/mol
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-621-534
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
T1374250ML
250 mL
Each of 1 for $1,032.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

13887-98-4
222.19 g/mol

Specifications

13887-98-4
222.19 g/mol
Approximately. 90%
3,6,9-Trioxaundecanedioic Acid
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.