Learn More
Fisher Science Education™ Benedict's Solution, Quantitative

Specifications
| Chemical Name or Material | Benedict's Solution, Quantitative |
|---|---|
| Color | Undesignated |
| CAS | 7758-99-8 , 63126-89-6 , 7732-18-5 |
| Assay Percent Range | <2% w/v |
| Density | 1.2g/mL |
Description
This product is for educational use - no C of A is available.
Specifications
| Benedict's Solution, Quantitative | |
| 7758-99-8 , 63126-89-6 , 7732-18-5 | |
| 1.2g/mL |
| Undesignated | |
| <2% w/v | |
| Solution |
Safety and Handling
ShelfLife : 2 years
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.