Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Glyceraldehyde, Spectrum™ Chemical
SDP

Synonym: Glyceraldehyde, 2,3-Dihydroxypropanal, Glyceric Aldehyde

$689.00 - $2910.00

Chemical Identifiers

CAS 56-82-6
Molecular Weight (g/mol) 90.08 g/mol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-620-787
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
G33075GM
5 g
Each for $689.00
Add to Cart
 
18-620-788
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
G330725GM
25 g
Each for $2,910.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

56-82-6
90.08 g/mol

Specifications

56-82-6
90% Min.
DL-Glyceraldehyde
90.08 g/mol
Bottle
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.