Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyceryl Tribenzoate, Spectrum™ Chemical
SDP

Synonym: Tribenzoin, Glycerol Tribenzoate, 1,2,3-Propanetriol Tribenzoate

$465.00 - $2479.00

Chemical Identifiers

CAS 614-33-5
Molecular Weight (g/mol) 404.41 g/mol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-620-771
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
G14761KG
1 kg
Each for $465.00
Add to Cart
 
18-620-772
SDP
missing translation for 'viewDocuments'
Spectrum Chemical Manufacturing Corporation
G147610KG
10 kg
Each for $2,479.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

614-33-5
404.41 g/mol

Specifications

614-33-5
Approximately 95%
Glyceryl Tribenzoate
404.41 g/mol
Amber Bottle
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.