Learn More
LiChropur™ Hydrogen Chloride Ethanol Solution, ∼1.25 M HCl, For GC Derivatization, MilliporeSigma™ Supelco™
$63.70 - $208.00
Specifications
| CAS | 7647-01-0 |
|---|---|
| Linear Formula | HCl · CH3CH2OH |
| MDL Number | MFCD00011324 |
| Density | 0.825 g/mL (at 20°C) |
| Synonym | Hydrochloric acid in ethanol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
11-102-0462
|
MilliporeSigma Supelco
1793450ML |
50 mL |
Each for $63.70
|
|
|||||
|
11-101-9026
|
MilliporeSigma Supelco
17934250ML |
250 mL |
Each for $208.00
|
|
|||||
Description
Specifications
| 7647-01-0 | |
| MFCD00011324 | |
| Hydrochloric acid in ethanol | |
| GC Derivatization |
| HCl · CH3CH2OH | |
| 0.825 g/mL (at 20°C) | |
| 36.46 |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.