Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marble’s Reagent, Reagents

Marble’s Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$83.00 - $126.00

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-334-743
Accessories View Documents
Reagents Holdings Llc
CM1150001C
1 L
Each for $126.00
Only null left
Add to Cart
 
 Restricted Item
01-334-742
Accessories View Documents
Reagents Holdings Llc
CM115000500C
500 mL
Each for $83.00
Only null left
Add to Cart
 
 Restricted Item
Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 7647-01-0
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Green
H2O
O
18.02
Glass Bottle
<1
Liquid
7732-18-5 , 7647-01-0
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.15
Non-sterile
Marble’s Reagent
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.