Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Methionine/Choline Deficient Diet (Powdered)

Methionine/Choline Deficient Diet (Powdered) has been used to study nonalcoholic steatohepatitis (NASH).

$1516.00 - $5220.00

Specifications

Format Powder
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
02-960-43810
View Documents
MP Biomedicals™
0296043810
10 kg
Each for $1,516.00
Add to Cart
 
02-960-43820
View Documents
MP Biomedicals™
0296043820
20 kg
Each for $2,455.00
Add to Cart
 
02-960-43850
View Documents
MP Biomedicals™
0296043850
50 kg
Each for $5,220.00
Add to Cart
 
Description

Description

  • Contains sucrose, corn starch, corn oil, Alphacel non-nutritive bulk, and AIN 76 mineral mix.
Specifications

Specifications

Powder
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.