Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Glycocholate, Spectrum™ Chemical
SDP

$174.00 - $65512.00

Chemical Identifiers

CAS 863-57-0
Molecular Weight (g/mol) 487.62 g/mol
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-621-485
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
S22761GM
1 g
Each for $174.00
Add to Cart
 
18-621-486
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
S22765GM
5 g
Each for $770.00
Add to Cart
 
18-621-487
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
S227625GM
25 g
Each for $3,030.00
Add to Cart
 
18-621-488
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
S227650GM
50 g
Each for $4,080.00
Add to Cart
 
18-621-489
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
S22765KGBL
5 kg
Each for $65,512.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

863-57-0
487.62 g/mol

Specifications

863-57-0
487.62 g/mol
Sodium Glycocholate
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.