Learn More
Fisher Science Education™ Sodium Silicate Solution, 37%

$13.84 - $45.40
Chemical Identifiers
| CAS | 1344-09-8 , 7732-18-5 |
|---|---|
| Molecular Formula | H10Na2O8Si |
| Molecular Weight (g/mol) | 212.14 |
| MDL Number | MFCD00003492 |
| InChI Key | ADPGKKZKGXANON-UHFFFAOYSA-N |
| Synonym | sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass |
| PubChem CID | 23266 |
| ChEBI | CHEBI:60720 |
| IUPAC Name | disodium oxosilanebis(olate) pentahydrate |
| SMILES | O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O |
| Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Quantity | Packaging | Price | Quantity & Availability | |||||
S25566
![]() |
500 mL | 37% |
Each for $13.84
Case of 20 EA for $230.00
|
|
|||||
S25566A
![]() |
1 L | 37% |
Each for $45.40
Case of 12 EA for $496.00
|
|
|||||
S25566B
![]() |
3.785 L | 37% | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
This product is for educational use - no C of A is available.
Chemical Identifiers
| 1344-09-8 , 7732-18-5 | |
| 212.14 | |
| ADPGKKZKGXANON-UHFFFAOYSA-N | |
| 23266 | |
| disodium oxosilanebis(olate) pentahydrate |
| H10Na2O8Si | |
| MFCD00003492 | |
| sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass | |
| CHEBI:60720 | |
| O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O |
Specifications
| Colorless | |
| 37% w/v | |
| MFCD00003492 | |
| ADPGKKZKGXANON-UHFFFAOYSA-N | |
| disodium oxosilanebis(olate) pentahydrate | |
| 23266 | |
| Solution |
| 1344-09-8 , 7732-18-5 | |
| H10Na2O8Si | |
| sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass | |
| O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O | |
| 212.14 | |
| CHEBI:60720 | |
| Sodium Silicate Solution, 37% |
Safety and Handling
ShelfLife : 5 years
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.