Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofolic Acid, Spectrum™ Chemical
SDP

$1867.00

Chemical Identifiers

CAS 135-16-0
Molecular Weight (g/mol) 445.43 g/mol
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-621-650
SDP
View Documents Promotion Details
Spectrum Chemical Manufacturing Corporation
T28241GM
1 g
Each of 1 for $1,867.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

135-16-0
445.43 g/mol

Specifications

135-16-0
445.43 g/mol
Amber Glass Bottle
Tetrahydrofolic Acid
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.