Assay Details
| Gene Symbol: LIG_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_317993.4 | XP_317993.4 | 1 - 2 | 55 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIG_ANOGA| Gene Symbol: LIG_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_317993.4 | XP_317993.4 | 1 - 2 | 55 | 66 | |
Set Membership: |
|