Assay Details
| Gene Symbol: CPR139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435683.1 | XP_003435731.1 | 2 - 3 | 195 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CPR139| Gene Symbol: CPR139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003435683.1 | XP_003435731.1 | 2 - 3 | 195 | 75 | |
Set Membership: |
|