Assay Details
| Gene Symbol: CDC42_ANOGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003436078.1 | XP_003436126.1 | 4 - 5 | 828 | 63 | |
| XM_003436079.1 | XP_003436127.1 | 4 - 5 | 927 | 63 | ||
| XM_312505.5 | XP_312505.3 | 4 - 5 | 755 | 63 | ||
More Information
Set Membership: |
|

CDC42_ANOGA
Genomic Map