Assay Details
| Gene Symbol: LOX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101603.3 | NP_564021.1 | 1 - 2 | 583 | 73 | |
| GenBank mRNA | AY075625.1 | - | 1 - 2 | 495 | 73 | |
| BT006348.1 | - | 1 - 2 | 394 | 73 | ||
More Information
Set Membership: |
|

LOX3
Genomic Map