Assay Details
| Gene Symbol: NF-YA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198095.1 | NP_001185024.1 | 4 - 5 | 933 | 66 | |
| NM_101621.3 | NP_173202.1 | 4 - 5 | 747 | 66 | ||
| NM_202121.2 | NP_973850.1 | 4 - 5 | 733 | 66 | ||
| NM_202122.2 | NP_973851.1 | 3 - 4 | 1089 | 66 | ||
| GenBank mRNA | AB493463.1 | - | 3 - 4 | 454 | 66 | |
| BT033080.1 | - | 3 - 4 | 454 | 66 | ||
More Information
Set Membership: |
|

NF-YA8
Genomic Map