Assay Details
| Gene Symbol: MP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332415.1 | NP_001321214.1 | 10 - 11 | 1175 | 116 | |
| NM_101840.3 | NP_173414.1 | 9 - 10 | 1709 | 116 | ||
| GenBank mRNA | AF334716.1 | - | 9 - 10 | 1025 | 116 | |
| BT002050.1 | - | 9 - 10 | 1499 | 116 | ||
| BT008805.1 | - | 9 - 10 | 1025 | 116 | ||
More Information
Set Membership: |
|

MP
Genomic Map