Assay Details
| Gene Symbol: TAF11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101855.1 | NP_173429.1 | 1 - 2 | 90 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAF11b| Gene Symbol: TAF11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101855.1 | NP_173429.1 | 1 - 2 | 90 | 82 | |
Set Membership: |
|