Assay Details
| Gene Symbol: FNR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084097.1 | NP_001077566.1 | 6 - 7 | 880 | 109 | |
| NM_001332422.1 | NP_001319047.1 | 5 - 6 | 1215 | 109 | ||
| NM_101857.6 | NP_173431.1 | 6 - 7 | 972 | 109 | ||
| GenBank mRNA | AK318722.1 | - | 5 - 6 | 1102 | 109 | |
| AY062739.1 | - | 6 - 7 | 764 | 109 | ||
| AY114663.1 | - | 6 - 7 | 746 | 109 | ||
More Information
Set Membership: |
|

FNR2
Genomic Map