Assay Details
| Gene Symbol: GAMMA CA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126049.4 | NP_569036.1 | 2 - 3 | 394 | 82 | |
| GenBank mRNA | AY045784.1 | - | 2 - 3 | 362 | 82 | |
| AY079385.1 | - | 2 - 3 | 254 | 82 | ||
| AY087379.1 | - | 2 - 3 | 389 | 82 | ||
More Information
Set Membership: |
|

GAMMA CA3
Genomic Map