Assay Details
| Gene Symbol: DGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126066.4 | NP_569038.1 | 12 - 13 | 1386 | 73 | |
| GenBank mRNA | AF428338.1 | - | 12 - 13 | 1304 | 73 | |
| AY150381.1 | - | 12 - 13 | 1263 | 73 | ||
More Information
Set Membership: |
|

DGL1
Genomic Map