Assay Details
| Gene Symbol: FH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126147.4 | NP_201548.1 | 2 - 3 | 1954 | 66 | |
| GenBank mRNA | AY337456.1 | - | 2 - 3 | 1792 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FH6| Gene Symbol: FH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126147.4 | NP_201548.1 | 2 - 3 | 1954 | 66 | |
| GenBank mRNA | AY337456.1 | - | 2 - 3 | 1792 | 66 | |
Set Membership: |
|