Assay Details
| Gene Symbol: FH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126147.4 | NP_201548.1 | 3 - 4 | 2193 | 149 | |
| GenBank mRNA | AY337456.1 | - | 3 - 4 | 2031 | 149 | |
More Information
Set Membership: |
|

FH6
Genomic Map