Assay Details
| Gene Symbol: STI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335120.1 | NP_001324706.1 | 4 - 5 | 3146 | 115 | |
| NM_126303.3 | NP_565285.1 | 4 - 5 | 3135 | 115 | ||
| GenBank mRNA | AF264023.1 | - | 4 - 5 | 3012 | 115 | |
More Information
Set Membership: |
|

STI
Genomic Map