Assay Details
| Gene Symbol: DRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035542.1 | NP_001030619.1 | 6 - 7 | 1346 | 71 | |
| NM_111020.3 | NP_566141.1 | 6 - 7 | 1389 | 71 | ||
| NM_180161.2 | NP_850492.1 | 6 - 7 | 1411 | 71 | ||
| NM_202477.2 | NP_974206.1 | 6 - 7 | 1338 | 71 | ||
| GenBank mRNA | AB010259.1 | - | 5 - 6 | 1144 | 71 | |
| AF428313.1 | - | 6 - 7 | 1242 | 71 | ||
| AY050375.1 | - | 6 - 7 | 1340 | 71 | ||
| BT009670.1 | - | 5 - 6 | 1137 | 71 | ||
More Information
Set Membership: |
|

DRH1
Genomic Map