Assay Details
| Gene Symbol: SOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111057.4 | NP_186840.1 | 2 - 3 | 351 | 70 | |
| GenBank mRNA | AB071965.1 | - | 2 - 3 | 351 | 70 | |
| AF360247.1 | - | 2 - 3 | 232 | 70 | ||
| AK226455.1 | - | 2 - 3 | 233 | 70 | ||
| AY133863.1 | - | 2 - 3 | 194 | 70 | ||
More Information
Set Membership: |
|

SOX
Genomic Map